Tiazolidina-2,4-diona ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

T1990-25G T1990-500G T1990-100G
TCIAT1990-25GEA 30 EUR
TCIAT1990-25G TCIAT1990-500G TCIAT1990-100G
Tiazolidina-2,4-diona ≥98.0% (by GC, titration analysis)
Tiazolidina-2,4-diona
Formula: C₃H₃NO₂S
Número MDL: MFCD00005478
Núm. CAS: 2295-31-0
EINECS: 218-941-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2e9eabb8e5780dbd8b8267bda3be5cd5eb93cb2&vid=c2e9eabb8e5780dbd8b8267bda3be5cd5eb93cb2&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 124.0 to 128.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR