Ácido (3R,4S,5R)-(-)-3,4,5-trihidroxiciclohex-1-enocarboxílico ≥97.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
(3R,4S,5R)-3,4,5-Trihydroxycyclohex-1-ene-1-carboxylic acid, Shikimic acid
S0038-1G
S0038-100MG
S0038-5G
TCIAS0038-1GEA
39
EUR
TCIAS0038-1G
TCIAS0038-100MG
TCIAS0038-5G
Ácido (3R,4S,5R)-(-)-3,4,5-trihidroxiciclohex-1-enocarboxílico ≥97.0% (by titrimetric analysis)
Ácido (3R,4S,5R)-(-)-3,4,5-trihidroxiciclohex-1-enocarboxílico
Formula:
C₇H₁₀O₅ MW: 174,15 g/mol |
Número MDL:
MFCD00066278 Núm. CAS: 138-59-0 EINECS: 205-334-2 Índice Merck: 14,08480 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 97.0 % |
Specific rotation [a]20/D | -175.0 to -185.0 deg (C=1, H2O) |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...