1,4-Dietinilbenceno ≥98.0% (por GC)

Proveedor: TCI

Sinónimos: p-Diethynylbenzene, 1,4-Diethylnylbenzene

D2151-1G D2151-5G
TCIAD2151-1GEA 44 EUR
TCIAD2151-1G TCIAD2151-5G
1,4-Dietinilbenceno ≥98.0% (por GC)
1,4-Dietinilbenceno
Formula: C₁₀H₆
MW: 126.16 g/mol
Número MDL: MFCD00078375
Núm. CAS: 935-14-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96d8b3abebde65f72d36c7cb553b74d3353544ee&vid=96d8b3abebde65f72d36c7cb553b74d3353544ee&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 92.0 to 97.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR