Ácido 2-naftiloxiacético ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

N0045-25G N0045-500G
TCIAN0045-25GEA 28 EUR
TCIAN0045-25G TCIAN0045-500G
Ácido 2-naftiloxiacético ≥98.0% (by GC, titration analysis)
Ácido 2-naftiloxiacético
Formula: C₁₂H₁₀O₃
Número MDL: MFCD00004066
Núm. CAS: 120-23-0
EINECS: 204-380-0
Índice Merck: 14,06397
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2bff172705f8950fcfa3bbad1c450766ff9f67bf&vid=2bff172705f8950fcfa3bbad1c450766ff9f67bf&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 155.0 to 158.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR