4-tert-Butilcalix[4]areno ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
Sinónimos:
p-tert-Butylcalix[4]arene
B1809-1G
B1809-25G
B1809-5G
TCIAB1809-1GEA
17
EUR
TCIAB1809-1G
TCIAB1809-25G
TCIAB1809-5G
4-tert-Butilcalix[4]areno ≥98.0% (by HPLC)
4-tert-Butilcalix[4]areno
Formula:
C₄₄H₅₆O₄ MW: 648,93 g/mol |
Número MDL:
MFCD00066280 Núm. CAS: 60705-62-6 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...