4-tert-Butilcalix[8]areno ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
B1750-25G
B1750-5G
TCIAB1750-25GEA
306
EUR
TCIAB1750-25G
TCIAB1750-5G
4-tert-Butilcalix[8]areno ≥98.0% (by HPLC)
4-tert-Butilcalix[8]areno
Formula:
C₈₈H₁₁₂O₈ MW: 1297,85 g/mol |
Número MDL:
MFCD00075392 Núm. CAS: 68971-82-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...