4,4''-Dinitrilo-p-terfenilo ≥97.0% (by HPLC, total nitrogen)
Proveedor: TCI
D3394-1G
D3394-5G
TCIAD3394-1GEA
24
EUR
TCIAD3394-1G
TCIAD3394-5G
4,4''-Dinitrilo-p-terfenilo ≥97.0% (by HPLC, total nitrogen)
4,4''-Dinitrilo-p-terfenilo
Formula:
C₁₈H₁₂N₂O₄ Pto. de fusión: 274°C |
Número MDL:
MFCD00051743 Núm. CAS: 3282-11-9 |
|
Resultados test especificaciones
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...