5-Oxo-D-prolina ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

P1354-25G P1354-5G
TCIAP1354-25GEA 55 EUR
TCIAP1354-25G TCIAP1354-5G
5-Oxo-D-prolina ≥98.0% (by titrimetric analysis)
5-Oxo-D-prolina
Formula: C₅H₇NO₃
Número MDL: MFCD00066212
Núm. CAS: 4042-36-8
EINECS: 223-735-0
Índice Merck: 14,08001
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=926a56527b9dc5df86df98f056aa1f2356d6ce7e&vid=926a56527b9dc5df86df98f056aa1f2356d6ce7e&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Optical purity (LC) min. 99.0 ee%
Purity (Neutralization titration) min. 98.0 %
Melting point 159.0 to 164.0 deg-C
Specific rotation [a]20/D +8.0 to +12.0 deg (C=5, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR