N,N,N',N'-Tetrametilbencidina ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

T0141-1G T0141-5G
TCIAT0141-1GEA 62 EUR
TCIAT0141-1G TCIAT0141-5G
N,N,N',N'-Tetrametilbencidina ≥98.0% (by GC, titration analysis)
N,N,N',N'-Tetrametilbencidina
Formula: C₁₆H₂₀N₂
Número MDL: MFCD00008310
Núm. CAS: 366-29-0
EINECS: 206-676-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c06488e9b85fb3cebd9899ef29cb2c476a16c2a7&vid=c06488e9b85fb3cebd9899ef29cb2c476a16c2a7&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 192.0 to 198.0 deg-C
Solubility in Chloroform almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR