2,5-Dibromo-3,4-dinitrotiofeno ≥98.0% (by HPLC)

Proveedor: TCI

D2371-1G D2371-5G
TCIAD2371-1GEA 55 EUR
TCIAD2371-1G TCIAD2371-5G
2,5-Dibromo-3,4-dinitrotiofeno ≥98.0% (by HPLC)
2,5-Dibromo-3,4-dinitrotiofeno
Formula: C₄Br₂N₂O₄S
MW: 331,94 g/mol
Número MDL: MFCD00015537
Núm. CAS: 52431-30-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0d433154f6ba76c26316dcc43f48cea1b462fc4&vid=e0d433154f6ba76c26316dcc43f48cea1b462fc4&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 135.0 to 139.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR