Sulfato de bis(2-metilisotiouronio) ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
M0442-25G
M0442-500G
M0442-100G
TCIAM0442-25GEA
16
EUR
TCIAM0442-25G
TCIAM0442-500G
TCIAM0442-100G
Sulfato de bis(2-metilisotiouronio) ≥98.0% (by titrimetric analysis)
Sulfato de bis(2-metilisotiouronio)
Formula:
C₄H₁₂N₄S₂·H₂SO₄ |
Número MDL:
MFCD00013055 Núm. CAS: 867-44-7 EINECS: 212-759-7 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...