Pteridina-2,4-diol ≥99.0% (by titrimetric analysis)

Proveedor: TCI

L0086-1G L0086-5G
TCIAL0086-1GEA 79 EUR
TCIAL0086-1G TCIAL0086-5G
Pteridina-2,4-diol ≥99.0% (by titrimetric analysis)
Pteridina-2,4-diol
Formula: C₆H₄N₄O₂
Número MDL: MFCD00066250
Núm. CAS: 487-21-8
EINECS: 207-652-7
Índice Merck: 14,05596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5352e2f0fc2c4116ce0f68569f86ba560c2ee712&vid=5352e2f0fc2c4116ce0f68569f86ba560c2ee712&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 99.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR