Guanosina ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

G0171-100G G0171-25G G0171-5G
TCIAG0171-100GEA 86 EUR
TCIAG0171-100G TCIAG0171-25G TCIAG0171-5G
Guanosina ≥98.0% (by HPLC, titration analysis)
Guanosina
Formula: C₁₀H₁₃N₅O₅
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00010182
Núm. CAS: 118-00-3
EINECS: 204-227-8
Índice Merck: 14,04566
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4edb1fbea8e30e86e72cff16ab729385ce0d79f5&vid=faaf1c10e997b8023bf60922b414c967b74725cc&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -72.0 to -80.0 deg (C=1, 1mol/L NaOH sol.)

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR