5-Bromo-2-metoxifenol ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
B3342-25G
B3342-5G
TCIAB3342-25GEA
52
EUR
TCIAB3342-25G
TCIAB3342-5G
5-Bromo-2-metoxifenol ≥98.0% (by GC, titration analysis)
5-Bromo-2-metoxifenol
Formula:
C₇H₇BrO₂ MW: 203,0300 g/mol Pto. Ebullición: 150°C |
Número MDL:
MFCD00673137 Núm. CAS: 37942-01-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 63.0 to 67.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...