(R)-(-)-N-(3,5-Dinitrobenzoyl)-α-phenylglycine ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: (R)-(-)-N-(3,5-Dinitrobenzoyl)-alpha-phenylglycine, (R)-(-)-N-(3,5-Dinitrobenzoyl) phenylglycine, (R)-2-(3,5-Dinitrobenzamido)-2-phenylacetic acid, (R)-(-)-N-(3,5-Dinitrobenzoyl)-α-phenylglycine

D1853-1G D1853-5G
TCIAD1853-1GEA 19 EUR
TCIAD1853-1G TCIAD1853-5G
(R)-(-)-N-(3,5-Dinitrobenzoyl)-α-phenylglycine ≥98.0% (by HPLC, titration analysis)
(R)-(-)-N-(3,5-Dinitrobenzoyl)-α-phenylglycine
Formula: C₁₅H₁₁N₃O₇
MW: 345,26 g/mol
Número MDL: MFCD00010134
Núm. CAS: 74927-72-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fee2c381e71213d31a7bfca9f06f9dd51be4c59d&vid=fee2c381e71213d31a7bfca9f06f9dd51be4c59d&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -99.0 to -105.0 deg (C=1,THF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR