2'-Deoxitimidina ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine
T0233-1G
T0233-25G
T0233-5G
TCIAT0233-1GEA
16
EUR
TCIAT0233-1G
TCIAT0233-25G
TCIAT0233-5G
2'-Deoxitimidina ≥98.0% (by HPLC, titration analysis)
Timidina
Formula:
C₁₀H₁₄N₂O₅ MW: 242,23 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00006537 Núm. CAS: 50-89-5 EINECS: 200-070-4 Índice Merck: 14,09397 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 186.0 to 189.0 deg-C |
Specific rotation [a]20/D | +32.0 to +34.0 deg (C=1, 1mol/L NaOH sol.) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...