Ácido dibenzoil-D-tartarico monohidrato ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: (2S,3S)-2,3-Bis(benzoyloxy)succinic acid hydrate

D1398-25G D1398-500G
TCIAD1398-25GEA 16 EUR
TCIAD1398-25G TCIAD1398-500G
Ácido dibenzoil-D-tartarico monohidrato ≥98.0% (by titrimetric analysis)
Ácido dibenzoil-D-tartarico monohidrato
Formula: C₁₈H₁₄O₈·1H₂O
MW: 376.33 g/mol (358.31 anhy)
Número MDL: MFCD00150722
Núm. CAS: 80822-15-7
EINECS: 241-097-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84e54c35800f62b20a14763546bf93742b8e3323&vid=84e54c35800f62b20a14763546bf93742b8e3323&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +107 to +111 deg (C=1, EtOH)
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR