(1S)-(-)-Camphanic acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: (1S,4R)-4,7,7-Trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carboxylic acid

C1021-1G C1021-5G
TCIAC1021-1GEA 24 EUR
TCIAC1021-1G TCIAC1021-5G
(1S)-(-)-Camphanic acid ≥98.0% (by titrimetric analysis)
(1S)-(-)-Camphanic acid
Formula: C₁₀H₁₄O₄
MW: 198,22 g/mol
Número MDL: MFCD00044948
Núm. CAS: 13429-83-9
EINECS: 236-552-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e58513947cf970426eb939c9069e3b4399a8ea9b&vid=e58513947cf970426eb939c9069e3b4399a8ea9b&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystaline
Purity (GC) min. 96.0 %
Optical purity (GC) min. 99.0 ee%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR