1,8-Naphthyridine ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
1,8-Diazanaphthalene, Pyrido[2,3-b]pyridine
N0464-1G
N0464-100MG
TCIAN0464-1GEA
268
EUR
TCIAN0464-1G
TCIAN0464-100MG
1,8-Naphthyridine ≥98.0% (por GC)
1,8-Naphthyridine
Formula:
C₈H₆N₂ MW: 130,1466 g/mol |
Número MDL:
MFCD00059751 Núm. CAS: 254-60-4 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Solubility in Methanol | very faint turbidity |
Melting point | 95.0 to 99.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...