4,4'-Dimetil-2,2'-bipiridilo ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
4,4'-Dimethyl-2,2'-bipyridine
B0852-1G
B0852-25G
B0852-5G
TCIAB0852-1GEA
16
EUR
TCIAB0852-1G
TCIAB0852-25G
TCIAB0852-5G
4,4'-Dimetil-2,2'-bipiridilo ≥98.0% (by GC, titration analysis)
4,4'-Dimetil-2,2'-bipiridilo
Formula:
C₁₂H₁₂N₂ MW: 184.24 g/mol |
Número MDL:
MFCD00006441 Núm. CAS: 1134-35-6 EINECS: 214-483-2 |
|
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 172.0 to 174.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...