Nicotinohidrazida ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Nicotinohydrazide, (3-Pyridylcarbonyl)hydrazine, Pyridine-3-carbohydrazide
N0087-25G
N0087-100G
TCIAN0087-25GEA
21
EUR
TCIAN0087-25G
TCIAN0087-100G
Nicotinohidrazida ≥98.0% (by GC, titration analysis)
Nicotinohidrazida
Formula:
C₆H₇N₃O MW: 137,14 g/mol |
Número MDL:
MFCD00006383 Núm. CAS: 553-53-7 EINECS: 209-041-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Bromination Method) | min. 98.0 % |
Melting point | 160.0 to 164.0 deg-C |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...