Cloruro de (4-clorobencil)trifenilfosfonio ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

C1581-25G C1581-5G
TCIAC1581-25GEA 73 EUR
TCIAC1581-25G TCIAC1581-5G
Cloruro de (4-clorobencil)trifenilfosfonio ≥98.0% (by HPLC, titration analysis)
Cloruro de (4-clorobencil)trifenilfosfonio
Formula: C₂₅H₂₁Cl₂P
MW: 423.32 g/mol
Número MDL: MFCD00041533
Núm. CAS: 1530-39-8
EINECS: 216-228-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a3c6fdb83afdd29b5c7ef437fee147e059f01a4&vid=5a3c6fdb83afdd29b5c7ef437fee147e059f01a4&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR