Piridina-2-acetonitrilo ≥98.0% (por GC)
Proveedor: TCI
|
Peligro
|
P1014-25G
P1014-100G
TCIAP1014-25GEA
65
EUR
TCIAP1014-25G
TCIAP1014-100G
Piridina-2-acetonitrilo ≥98.0% (por GC)
Piridina-2-acetonitrilo
Formula:
C₇H₆N₂ |
Número MDL:
MFCD00006346 Núm. CAS: 2739-97-1 EINECS: 220-364-6 UN: 3439 ADR: 6.1,II |
|
Resultados test especificaciones
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...