5-Bromo-2'-deoxyuridina (Broxuridina) ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: 5-Bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Bromo-2'-deoxyuridine, 5-BrdU

B1575-1G B1575-5G
TCIAB1575-1GEA 32 EUR
TCIAB1575-1G TCIAB1575-5G
5-Bromo-2'-deoxyuridina (Broxuridina) ≥98.0% (by HPLC, titration analysis)
5-Bromo-2'-deoxyuridina (Broxuridina)
Formula: C₉H₁₁BrN₂O₅
MW: 307.10 g/mol
Número MDL: MFCD00006529
Núm. CAS: 59-14-3
EINECS: 200-415-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f60c43d1aef98eea7763606f5a7990b29b3b746b&vid=f60c43d1aef98eea7763606f5a7990b29b3b746b&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +20.0 to +24.0 deg (C=1, H2O)
Solubility in hot Water within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR