2-Mercaptobenzoxazol ≥98.0% (by GC, titration analysis)

Proveedor: TCI

Sinónimos: Benzo[d]oxazole-2-thiol

B0096-25G B0096-500G
TCIAB0096-25GEA 32 EUR
TCIAB0096-25G TCIAB0096-500G
2-Mercaptobenzoxazol ≥98.0% (by GC, titration analysis)
2-Mercaptobenzoxazole
Formula: C₇H₅NOS
MW: 151,19 g/mol
Número MDL: MFCD00005769
Núm. CAS: 2382-96-9
EINECS: 219-191-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f698d4b44b756252c988815952af2816e13bcf4c&vid=f698d4b44b756252c988815952af2816e13bcf4c&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 192.0 to 198.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR