2,2'-[(1,1-Dimetiletil)imino]bisetanol ≥97.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
2,2'-(tert-Butylimino)diethanol, N-Bis(2-hydroxyethyl)-tert-butylamine, N-tert-Butyldiethanolamine, 2,2'-(tert-Butylazanediyl)diethanol
B1194-25ML
B1194-500ML
TCIAB1194-25MLEA
16
EUR
TCIAB1194-25ML
TCIAB1194-500ML
2,2'-[(1,1-Dimetiletil)imino]bisetanol ≥97.0% (by titrimetric analysis)
2,2'-[(1,1-Dimetiletil)imino]bisetanol
Formula:
C₈H₁₉NO₂ Pto. Ebullición: 161°C |
Número MDL:
MFCD00020584 Núm. CAS: 2160-93-2 EINECS: 218-480-7 UN: 3259 ADR: 8,III |
|
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to lump |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 47.0 to 51.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...