6-Amino-1,3-dimethyluracil ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
6-Amino-1,3-dimethylpyrimidine-2,4(1H,3H)-dione, 4-Amino-1,3-Dimethyluracil
A1420-25G
A1420-500G
TCIAA1420-25GEA
18
EUR
TCIAA1420-25G
TCIAA1420-500G
6-Amino-1,3-dimethyluracil ≥98.0% (by HPLC, titration analysis)
6-Amino-1,3-dimethyluracil
Formula:
C₆H₉N₃O₂ MW: 155,1546 g/mol |
Número MDL:
MFCD00006552 Núm. CAS: 6642-31-5 EINECS: 229-662-0 |
Resultados test especificaciones
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in hot 1mol/L NaOH | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...