Tetrafluoroterephthalic acid ≥97.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: 2,3,5,6-Tetrafluoroterephthalic acid

T0930-1G T0930-5G
TCIAT0930-1GEA 36 EUR
TCIAT0930-1G TCIAT0930-5G
Tetrafluoroterephthalic acid ≥97.0% (by titrimetric analysis)
Tetrafluoroterephthalic acid
Formula: C₈H₂F₄O₄
Número MDL: MFCD00012301
Núm. CAS: 652-36-8
EINECS: 211-489-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62329ccac1d6960c79e19466441ac8bdf9aa6162&vid=62329ccac1d6960c79e19466441ac8bdf9aa6162&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 97.0 %
Solubility in hot Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR