Tetrabutylammonium diiodoaurate
Proveedor: TCI
T1357-1G
T1357-100MG
TCIAT1357-1GEA
374
EUR
TCIAT1357-1G
TCIAT1357-100MG
Tetrabutylammonium diiodoaurate
Tetrabutylammonium diiodoaurate
Formula:
C₁₆H₃₆AuI₂N MW: 693,25 g/mol |
Número MDL:
MFCD00145387 Núm. CAS: 50481-03-3 |
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...