2-Amino-5-bromopyrazine ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

A1683-1G A1683-5G
TCIAA1683-1GEA 43 EUR
TCIAA1683-1G TCIAA1683-5G
2-Amino-5-bromopyrazine ≥98.0% (by GC, titration analysis)
2-Amino-5-bromopyrazine
Formula: C₄H₄BrN₃
Número MDL: MFCD00235015
Núm. CAS: 59489-71-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0230bb8dcd59e5d9c343da9cb0746588c69a1e2f&vid=0230bb8dcd59e5d9c343da9cb0746588c69a1e2f&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 112.0 to 116.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR