N-Acetyl-DL-methionine ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: Ac-DL-Met-OH

A0100-25G A0100-500G
TCIAA0100-25GEA 16 EUR
TCIAA0100-25G TCIAA0100-500G
N-Acetyl-DL-methionine ≥98.0% (by titrimetric analysis)
N-Acetyl-DL-methionine
Formula: C₇H₁₃NO₃S
Número MDL: MFCD00008681
Núm. CAS: 1115-47-5
EINECS: 214-224-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7edb33a53efd928a09b409a4d0fbc32212d59fa2&vid=7edb33a53efd928a09b409a4d0fbc32212d59fa2&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 113.0 to 117.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR