Ácido aminoiminometanosulfinico ≥93.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
Thiourea dioxide, FormamidinesulphInic acid
F0115-25G
F0115-500G
TCIAF0115-25GEA
16
EUR
TCIAF0115-25G
TCIAF0115-500G
Ácido aminoiminometanosulfinico ≥93.0% (by titrimetric analysis)
Ácido aminoiminometanosulfinico
Formula:
CH₄N₂O₂S MW: 108,12 g/mol |
Número MDL:
MFCD00002397 Núm. CAS: 1758-73-2 EINECS: 217-157-8 UN: 3341 ADR: 4.2,III |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Bromination Method) | min. 93.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...