2-Amino-5-clorobenzamida ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

A1970-1G A1970-5G
TCIAA1970-1GEA 18 EUR
TCIAA1970-1G TCIAA1970-5G
2-Amino-5-clorobenzamida ≥98.0% (by HPLC, titration analysis)
2-Amino-5-clorobenzamida
Formula: C₇H₇ClN₂O
Número MDL: MFCD00017126
Núm. CAS: 5202-85-7
EINECS: 225-990-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ea3e2578b098bd56bf310c9428da78fd5365ae7&vid=2ea3e2578b098bd56bf310c9428da78fd5365ae7&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 169.0 to 173.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR