Bitionol ≥97.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

T0865-25G T0865-500G
TCIAT0865-25GEA 24 EUR
TCIAT0865-25G TCIAT0865-500G
Bitionol ≥97.0% (by GC, titration analysis)
Bitionol
Formula: C₁₂H₆Cl₄O₂S
Número MDL: MFCD00055727
Núm. CAS: 97-18-7
EINECS: 202-565-0
UN: 2811
ADR: 6.1,II
Índice Merck: 14,01306
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b78f356cc81a9305bf903cf3c520fd8db3bbe996&vid=b78f356cc81a9305bf903cf3c520fd8db3bbe996&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 184.0 to 188.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR