Sulfato de 2-metil-p-fenilendiamina ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

D1602-500G D1602-25G
TCIAD1602-500GEA 244 EUR
TCIAD1602-500G TCIAD1602-25G
Sulfato de 2-metil-p-fenilendiamina ≥98.0% (by HPLC, titration analysis)
Sulfato de 2-metil-p-fenilendiamina
Formula: C₇H₁₂N₂O₄S
Número MDL: MFCD00013003
Núm. CAS: 615-50-9
EINECS: 210-431-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79e4c82306ae295b16d05b7040e92f6589387481&vid=79e4c82306ae295b16d05b7040e92f6589387481&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR