Tris(trimethylsilyl)phosphite ≥95.0% (by NMR)
Proveedor: TCI
|
Atención
|
Sinónimos:
Tris(trimethylsilyl)phosphite
P1217-25ML
P1217-5ML
TCIAP1217-25MLEA
214
EUR
TCIAP1217-25ML
TCIAP1217-5ML
Tris(trimethylsilyl)phosphite ≥95.0% (by NMR)
Tris(trimethylsilyl)phosphite
Formula:
C₉H₂₇O₃PSi₃ MW: 298,54 g/mol Pto. Ebullición: 73°C |
Número MDL:
MFCD00015588 Núm. CAS: 1795-31-9 |
Resultados test especificaciones
Appearance | Colorless to Almost colorless clear liquid |
Purity (Neutralization titration) | min. 97.0 % |
Purity (NMR) | min. 95.0 w/w% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...