(R)-(+)-Menthyltoluene-4-sulphinate ≥97.0% (by HPLC)

Proveedor: TCI

M1066-1G M1066-5G
TCIAM1066-1GEA 37 EUR
TCIAM1066-1G TCIAM1066-5G
(R)-(+)-Menthyltoluene-4-sulphinate ≥97.0% (by HPLC)
(R)-(+)-Menthyltoluene-4-sulphinate
Formula: C₁₇H₂₆O₂S
Temperatura de almacenaje: Congelador
Número MDL: MFCD00064472
Núm. CAS: 91796-57-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba4772ccd0a53123e8e18dcb7f9f11b2786e2b53&vid=ba4772ccd0a53123e8e18dcb7f9f11b2786e2b53&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 102.0 to 106.0 deg-C
Specific rotation [a]20/D +195 to +205 deg (C=2, Acetone)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR