α,α-Diphenyl-N-methyl-L-prolinol ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
(S)-Alpha,Alpha-Diphenylmethylprolinol
H0768-1G
H0768-100MG
H0768-5G
TCIAH0768-1GEA
171
EUR
TCIAH0768-1G
TCIAH0768-100MG
TCIAH0768-5G
α,α-Diphenyl-N-methyl-L-prolinol ≥98.0% (por GC)
α,α-Diphenyl-N-methyl-L-prolinol
Formula:
C₁₈H₂₁NO MW: 267,365 g/mol |
Número MDL:
MFCD00145245 Núm. CAS: 110529-22-1 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystaline |
Purity (GC) | min. 98.0 % |
Melting point | 68.0 to 72.0 deg-C |
Specific rotation [a]20/D | +52.0 to +57.0 deg (C=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...