2,4'-Metilendifenol ≥98.0% (por GC)

Proveedor: TCI
Atención

D1940-1G D1940-5G
TCIAD1940-1GEA 86 EUR
TCIAD1940-1G TCIAD1940-5G
2,4'-Metilendifenol ≥98.0% (por GC)
2,4'-Metilendifenol
Formula: C₁₃H₁₂O₂
Número MDL: MFCD00060113
Núm. CAS: 2467-03-0
EINECS: 219-579-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc90bc1faabe6526fc5a781ecf1f31efb28b1cef&vid=fc90bc1faabe6526fc5a781ecf1f31efb28b1cef&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 118.0 to 121.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR