2-Naftilacetonitrilo ≥98.0% (por GC)
Proveedor: TCI
|
Peligro
|
N0874-25G
N0874-5G
TCIAN0874-25GEA
181
EUR
TCIAN0874-25G
TCIAN0874-5G
2-Naftilacetonitrilo ≥98.0% (por GC)
2-Naftilacetonitrilo
Formula:
C₁₂H₉N MW: 167,21 g/mol |
Número MDL:
MFCD00004122 Núm. CAS: 7498-57-9 EINECS: 231-352-5 UN: 3439 ADR: 6.1,III |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 84.0 to 88.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...