4,5-Dimetiltiazol-2-amina monoclorhidrato ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
4,5-Dimethylthiazol-2-amine hydrochloride
A0292-1G
A0292-5G
TCIAA0292-1GEA
34
EUR
TCIAA0292-1G
TCIAA0292-5G
4,5-Dimetiltiazol-2-amina monoclorhidrato ≥98.0% (by titrimetric analysis)
4,5-Dimetiltiazol-2-amina monoclorhidrato
Formula:
C₅H₉ClN₂S MW: 164.66 g/mol |
Número MDL:
MFCD00012711 Núm. CAS: 71574-33-9 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...