4-Acetamidobenzenesulphonyl azide ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
A1786-25G
A1786-5G
A1786-100G
TCIAA1786-25GEA
79
EUR
TCIAA1786-25G
TCIAA1786-5G
TCIAA1786-100G
4-Acetamidobenzenesulphonyl azide ≥98.0% (by HPLC)
4-Acetamidobenzenesulphonyl azide
Formula:
C₈H₈N₄O₃S MW: 240,24 g/mol |
Número MDL:
MFCD00029626 Núm. CAS: 2158-14-7 |
Resultados test especificaciones
Appearance | White to Amber to Dark purple powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 110.0 to 114.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...