Ácido 6-bromo-2-naftoico ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
B3823-25G
B3823-5G
TCIAB3823-25GEA
165
EUR
TCIAB3823-25G
TCIAB3823-5G
Ácido 6-bromo-2-naftoico ≥98.0% (by GC, titration analysis)
Ácido 6-bromo-2-naftoico
Formula:
C₁₁H₇BrO₂ MW: 251,08 g/mol |
Número MDL:
MFCD01075720 Núm. CAS: 5773-80-8 |
|
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
![](/m-es.vwr.com/es_ES/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...