2,2-Dimetil-1,3-dioxano-4,6-diona ≥98.0% (by titrimetric analysis)

Proveedor: TCI

M0799-100G M0799-25G M0799-500G
TCIAM0799-100GEA 55 EUR
TCIAM0799-100G TCIAM0799-25G TCIAM0799-500G
2,2-Dimetil-1,3-dioxano-4,6-diona ≥98.0% (by titrimetric analysis)
2,2-Dimetil-1,3-dioxano-4,6-diona
Formula: C₆H₈O₄
Temperatura de almacenaje: Nevera
Número MDL: MFCD00006638
Núm. CAS: 2033-24-1
EINECS: 217-992-8
Índice Merck: 14,05817
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d9d4119601930e5344d5cdc9a719a33b6195f23&vid=4d9d4119601930e5344d5cdc9a719a33b6195f23&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Dark green powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 94.0 to 97.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR