Menadiona ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

M0373-25G M0373-250G
TCIAM0373-25GEA 44 EUR
TCIAM0373-25G TCIAM0373-250G
Menadiona ≥98.0% (by HPLC, titration analysis)
Menadiona
Formula: C₁₁H₈O₂
Número MDL: MFCD00001681
Núm. CAS: 58-27-5
EINECS: 200-372-6
Índice Merck: 14,05831
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc8a679d9709d8c4b101980f066a893080244425&vid=fc8a679d9709d8c4b101980f066a893080244425&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Cerium redox Method) min. 98.0 %
Melting point 105.0 to 108.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR