Solid phase extraction cartridges, Bond Elut™ C18
Proveedor: VARIAN
12102001 12102028 12113001 12113027 12102025 12102099 12162028B 12166001B 75401050 12102058 12102052 12113024 14102025 14102028 14256031 22102001 7540101C 7550101C 14256023 12256060 14113024 14113027 14256015 14256001 14102001 14102058 14113001 14102052 12105027 12102118 12102096 75502050 75501025 75501050 75401025 12102052T 14102028B 12102058B 52102001 52102024 52102052 52113024 12105102B
551-0300EA 0 EUR
551-0300 551-0315 551-0343 551-0345 551-0312 636-2504 VARI12162028B 551-0380 551-0425 551-0328 551-0327 VARI12113024 549-0844 549-0845 549-0869 VARI22102001 551-0424 VARI7550101C 549-0865 549-0836 VARI14113024 VARI14113027 549-0860 549-0855 549-0840 549-0850 VARI14113001 549-0849 549-0806 549-0802 549-0796 VARI75502050 VARI75501025 VARI75501050 VARI75401025 549-0785 VARI14102028B 549-0787 VARI52102001 VARI52102024 VARI52102052 VARI52113024 VARI12105102B
Solid phase extraction cartridges, Bond Elut™ C18
Columnas de cromatografía
Bond Elut™ C18 is the most hydrophobic, bonded silica sorbent in the Bond Elut™ range. It is the most popular SPE sorbent because of its extremely retentive nature for non-polar compounds. C18 is generally regarded as having the broadest spectrum of retention among bonded silica sorbents, since it retains most organic analytes from aqueous matrices. When analysing small to intermediate molecules, Bond Elut™ C18 can be used for desalting aqueous matrices prior to ion exchange, as salts pass through the sorbent unretained.
  • The most hydrophobic, bonded silica sorbent
  • Extremely retentive
More Product Information
Order Now
ESPECIFICACIONES
Environmentally Preferable
Carga de carbono 17,4% 17,4% 17,4%
Extremo tapado Yes Yes Yes
Surface area 50 m²/g 50 m²/g 50 m²/g