Microorganisms, EZ-CFU™ One Step
Proveedor: MICROBIOLOGICS
0392Z 0443Z 0483Z 0485Z 0486Z 0487Z 0484Z 0363Z 0688Z
0392ZEA 0 EUR
0392Z 0443Z 0483Z 0485Z 0486Z 0487Z 0484Z 0363Z 0688Z
Microorganisms, EZ-CFU™ One Step
Tests para microbiología Tests de microorganismos
The Growth Promotion Test (GPT) is an important function in the United States (USP), European (Ph. Eur.) and Japanese (JP) Pharmacopeias. EZ-CFU™ One Step quantitative microorganism preparations are used to perform Growth Promotion Tests of culture media with ease. No dilutions are required. EZ-CFU™ One Step is designed to deliver 10 to 100 CFU with each 0,1 ml inoculum. Each 2 ml vial of hydrated suspension offers 19 inocula. EZ-CFU™ One Step can be used for up to eight hours after hydration if the suspension is refrigerated between uses*. For added convenience, a peel-off Certificate of Assay is included for quality control documentation. The ATCC licensed derivative emblem, the ATCC licensed derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.
  • Kit contains microorganism and hydrating fluid
  • No dilutions required
  • Quantitative
  • Does not require frozen storage conditions
Información de suministro: EZ-CFU™ One Step kits include: 20 lyophilized pellets of a single quantitative microorganism, 10 vials of hydrating fluid (2 ml in each vial), Peel-Off Certificate of Assay.
Attention: Other microorganisms are available in the same format. Please contact Avantor to get more information.