D(-)-allo-Isoleucina ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: (2R,3S)-(-)-allo-Isoleucina, H-D-allo-Ile-OH

A0212-1G A0212-5G A0212-25G
TCIAA0212-1GEA 29 EUR
TCIAA0212-1G TCIAA0212-5G TCIAA0212-25G
D(-)-allo-Isoleucina ≥98.0% (by titrimetric analysis)
D(-)-allo-Isoleucina
Formula: C₆H₁₃NO₂
Número MDL: MFCD00066445
Núm. CAS: 1509-35-9
EINECS: 216-143-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=765f3959a3b6c20cff1b47b58b6bd7b69315e442&vid=765f3959a3b6c20cff1b47b58b6bd7b69315e442&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -35.0 to -39.0 deg (C=4, 6mol/L HCl)
Solubility in HCl (1+1) very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR