Ácido (2,3-difluorofenil)metabórico (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
Dihydroxy(2,3-difluorophenyl)borane, 2,3-Difluorobenzeneboronic acid, (2,3-Difluorophenyl)metaboric acid
D3523-25G
D3523-5G
TCIAD3523-25GEA
137
EUR
TCIAD3523-25G
TCIAD3523-5G
Ácido (2,3-difluorofenil)metabórico (contains varying amounts of Anhydride)
Ácido (2,3-difluorofenil)metabórico
Formula:
C₆H₅BF₂O₂ MW: 157.91 g/mol |
Número MDL:
MFCD01863170 Núm. CAS: 121219-16-7 |
|
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 113.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...