Ácido (2,5-diclorofenil)metabórico (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

D3044-25G D3044-5G
TCIAD3044-25GEA 196 EUR
TCIAD3044-25G TCIAD3044-5G
Ácido (2,5-diclorofenil)metabórico (contains varying amounts of Anhydride)
Ácido (2,5-diclorofenil)metabórico
Formula: C₆H₅BCl₂O₂
MW: 190,82 g/mol
Número MDL: MFCD01863182
Núm. CAS: 135145-90-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9daa03dcfd3ba185f1d0dc05a9c49ea1c605bb5e&vid=9daa03dcfd3ba185f1d0dc05a9c49ea1c605bb5e&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) 97.0 to 111.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR